Order Entry
Mexico
Orders LinkContactUsLinkComponent
VWR®, Beaker Cups, Polystyrene
VWR®, Beaker Cups, Polystyrene
20 ml (0.68 oz.)
Catalog # 13916-004
Supplier:   VWR International
VWR®, Beaker Cups, Polystyrene
Catalog # 13916-004
Supplier:  VWR International
VWR®, Beaker Cups, Polystyrene
VWR®, Beaker Cups, Polystyrene
20 ml (0.68 oz.)
Price
$639.30
Log in for availability
Case (500UN)
Product Details & Documents
Beaker cups are designed for use with various automated procedures and as an economical, general-purpose sample cup.
  • Disposable
  • Polystyrene fabrication
  • Economical sample cup
The 5 mL and 20 mL sizes are recommended for flame photometry analysis.
Specifications
  • Capacity:
    20 ml (0.68 oz.)