Order Entry
Mexico
ContactUsLinkComponent
 
Clay Pebbles for Hydroponics
  470004-346
undefined
Clay Pebbles for Hydroponics
  470004-346
 :  GMC10L

 

  • Description:
    Clay pebbles

 

 

Support and biofilter.

  • Extremely stable in both pH and EC

  • Made from 100% natural clay
  • Helps to prevent dripper systems from getting blocked
  • Pre-washed to aid in stability
  • Pebble size: 4 to 16 mm

Horticultural clay pebbles are made from 100% natural clay. They are clean, pH stable, and offer great aeration and drainage in hydroponics, especially in flood and drain, deep water culture, and drip feed systems. Due to their unique structure and ability to cover a large surface area, they offer the ideal environment to foster beneficial bacterial growth around the root zone, leading to naturally healthier plants.