Order Entry
Mexico
ContactUsLinkComponent
Chloramine T (sodium salt) trihydrate ≥98.0% (by titrimetric analysis)
Corrosivo
Salud
Irritante
Chloramine T (sodium salt) trihydrate ≥98.0% (by titrimetric analysis)
Catalog # TCC0076-25G
Proveedor:  TCI America
Número CAS:  
Corrosivo
Salud
Irritante
Chloramine T (sodium salt) trihydrate ≥98.0% (by titrimetric analysis)
Catalog # TCC0076-25G
Proveedor:  TCI America
Supplier Number:  C0076-25G
Número CAS:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Especificaciones

  • Size:
    25 g
  • Cat. no.:
    TCC0076-25G
  • Supplier No.:
    C0076-25G

Especificaciones

Resultados de prueba de especificación

  • Purity/Analysis Method:
    98.0% (T)

Resultados de prueba de especificación

Acerca de este artículo

Número CAS:7080-50-4
 :
Fórmula:CH₃C₆H₄SO₂NClNa·3H₂O
Número MDL:MFCD00149066
Índice Merck:13,02085
Punto de fusión:167…170 °C
Punto de fusión:167 - 170 °C
Densidad:1.43 g/cm³ (20 °C)
Punto de inflamación:192 °C
Punto de inflamación:192 °C
Temperatura de almacenamiento:Ambient
EINECS:204-854-7
ADR:8,III
 :281.69 g/mol
UN:3263
 :GHS05
 :GHS08
 :GHS07