Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Especificaciones
- Description:Sporicidin® Enzymatic cleaner concentrate
- Material:Cellulose/protein enzymes
- Size:3.8 L (1 gal.)
- Cat. no.:21909-840
- Supplier No.:ENZ-1284
Especificaciones
Acerca de este artículo
Sporicidin® enzymatic cleaner is a blend of enzymes in a low foaming solution formulated to loosen and remove dried debris, protein, blood, and mucin deposits. It effectively cleans both natural and synthetic fibers that have been soiled with organic and inorganic debris.
- Special blend contains cellulose and protein enzymes for cleaning and removing material stains
- Breaks down both organic and inorganic debris
- Mildly alkaline (pH 7.5) and contains enzyme preservative for added stability
- Low foaming, concentrated formulation
- Safe for use on rubber, stainless steel, vinyl, plastics, linens, clothing, carpets, fabrics and upholstery
- Reduces the need for manual scrubbing
- Non-flammable, non-staining, and non-toxic
Sporicidin® Enzymatic Cleaner can be used in ultrasonic units, automatic washers, and evacuation systems. This highly effective cleaner and stain remover has a neutral pH and is biodegradable, nonflammable and nontoxic. It is ideal for household, medical, dental, institutional, healthcare, veterinary, and cleanroom use.
Economical, highly concentrated formula:
1 ounce makes 1 gallon of cleaning solution
1 gallon (3.8 l) of concentrate makes 128 gallons (484.5 l) of solution
55 gallons (208.2 l) of concentrate makes 7040 gallons (26649 l) of solution