About this item
Cleans, disinfects, deodorizes. Provides 100% kill of pathogenic vegetative organisms, including MRSA, VRE and Avian Influenza A Virus (H9N2 and H1N1). Continuous residual activity up to 6 months. Compatible with plastics, wood, glass and metals, alcohol-free Sporicidin Disinfectant is non-staining, non-abrasive and non-corrosive. EPA and FDA registered; OSHA compliant. Lowest EPA toxicity category.
- EPA-registered bactericide, fungicide, virucide (both enveloped and non-enveloped viruses) and tuberculocide
- Disinfects, cleans and deodorizes
- Provides residual protection against odor-causing organisms and fungi
Invented by the dentist that created Chloraseptic® Sore Throat Spray, Sporicidin® disinfectants utilize the same active ingredient system. Sporicidin® disinfectants not only kill disease-causing microbes, they also continue to keep odor-causing bacteria away for as long as six months, if not removed from the surface.
When it comes to safety, Sporicidin® disinfectants have EPA toxicity ratings of Category IV – Very Low acute oral, acute dermal, acute inhalation, and skin irritation. Category IV – Very Low Toxicity is the lowest toxicity rating given to antimicrobials. Whether cleaning up a trauma scene, a MRSA contamination, or a simple sewage backflow, Sporicidin® disinfectant products are the right choice.