

Sulphuric acid fuming approx. 20% free SO₃ 20%
Catalog # AA45543-57
Supplier: Avantor
CAS Number:
Specifications
- Size:4 x 500 g
- Cat. no.:AA45543-57
- Supplier No.:45543-57
Specifications
About this item
:
Formula:H₂O₇S₂
MDL Number:MFCD00137792
Merck Index:14,8974
Density:1.9
Storage Temperature:Ambient
UN:1831
ADR:8,I
:80.06
:GHS05
:GHS07
CAS Number:8014-95-7
Formula:H₂O₇S₂ (H₂SO₄ + SO₃)