To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Size:500 g
- Environmentally Preferable:
- Cat. no.:71002-338
- Supplier No.:1.00043.0500
Specifications
About this item
For the enumeration of Bifidobacteria in dairy products. 62.5g in 950mL of purified water.
Contents: peptone from casein 10.0; Yeast extract 1.0; potassium di-hydrogen phosphate 3.0; Di-potassium hydrogen phosphate 4.8; Ammonium sulfate 3.0; Magnesium sulfate heptahydrate 0.2; L-cysteine monochloride 0.5; Sodium Propionate 15.0; Galactooligosaccharide(TOS) 10.0; Agar Agar 15.0.
For the enumeration of Bifidobacteria in dairy products
62.5g in 950mL of purified water