Order Entry
Mexico
Orders LinkContactUsLinkComponent
NeuLog pH Logger Sensor

NeuLog pH Logger Sensor

NeuLog pH logger sensor, pH: 0 - 14, pH: 0.01 pH
ADC: 16 bit
Catalog # 470137-380
Supplier #:  NUL206
NeuLog pH Logger Sensor
Catalog # 470137-380
NeuLog pH Logger Sensor
NeuLog pH Logger Sensor

NeuLog pH logger sensor, pH: 0 - 14, pH: 0.01 pH
ADC: 16 bit

Clearance
$122.20
Sale

Valid until 31/12/2026

Log in for availability
Each (1UN)
Product Details & Documents

This sensor can measure the static pH values of common liquids.

  • Durable electrode
  • Designed for long life
  • Sealed reference system
  • Gel fill
  • Maximum sample rate of 100 S/s

It also measures changing values in titrations or experiments. The sensor gives a fast response across the full pH range and can be calibrated with any standard buffer solution. Its sealed reference system and gel fill make it easy to use and maintain. With an epoxy body, it's a durable electrode to use in the laboratory and in the field.

Insert the sensor into a buffer of your choice, hold the push button for three seconds, and the reading will calibrate to that buffer's pH. It supports experiments lasting between one second and 31 days.

Ordering information: This unit construction features an epoxy body, round bulb, ASG VIII pH glass, gel-filled single junction reference with fiber frit, and Ag/AgCl wire. It requires a connection to a voltage source (a PC or NeuLog's display or battery unit).

Caution: For educational use only.

Specifications
  • Resolution:
    pH: 0.01 pH
    ADC: 16 bit
  • Description:
    NeuLog pH logger sensor
  • Range:
    pH: 0 - 14
  • Cat. No.:
    470137-380
  • Response time:
    30 s to 98% of full-scale at 25 °C
  • W×D×H:
    Electrode: 12 mm dia.; body: 150 mm; Cap: 16 mm O.D.
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • NeuLog USB module
    NeuLog USB module
    470137-442
    Log in for availability
    Each (1UN) - $2,059.56

Product Information

  • Catalog #
    Availability
    Price
  • NeuLog viewer graphic color display module
    NeuLog viewer graphic color display module
    470137-438
    Log in for availability
    Each (1UN) - $6,546.41

Product Information

  • Catalog #
    Availability
    Price
  • Neulog WiFi 2.0
    Neulog WiFi 2.0
    470221-194
    Log in for availability
    Each (1UN) - $174.60