NeuLog Soil Moisture Logger Sensor
ADC: 10 bit
Product Details & Documents
A unique approach to measuring moisture via vacuum pressure in a tensiometer.
- Max. sample rate 100 S/sec.
- Experimental duration 1 sec. to 31 days
The tensiometer on this logger sensor measures soil moisture via pressure level. When the soil is dry the water exits ceramic holes in the end due to diffusion, when the soil is wet the vacuum in the tensiometer pulls water inside and the vacuum decreases.
Ordering information: For each probe you will need one or more of the following items: Battery Module, Viewer module (not needed if connecting to a computer to display data), USB module (not needed if only using as a stand alone probe or you have the WIFI module), WIFI module (Not needed if connecting to a computer through the USB module). Recommended is to have a battery module and a USB module.
Caution: For educational use only.
Documents
Specifications
- Resolution:0.02 Kpa
ADC: 10 bit - Description:NeuLog soil moisture logger sensor
- Range:0 - 50 Kpa
- Cat. No.:470137-424
- Interface:USB
Accessories
Product Information
- Catalog #AvailabilityPrice
- NeuLog USB module
Product Information
- Catalog #AvailabilityPrice
- Neulog WiFi 2.0
Product Information
- Catalog #AvailabilityPrice
- NeuLog viewer graphic color display moduleNeuLog viewer graphic color display module470137-438Log in for availabilityEach (1UN) - $6,546.41