NeuLog Surface Temperature Logger Sensor
NeuLog surface temperature logger sensor, −40...+140 °C, 0.1 °C
ADC: 12 bit
ADC: 12 bit
470137-432
: NUL233
This tough sensor is usable on any surface or even in water.
- Two modes of operation: celsius and fahrenheit
- Sensor works on any surface
- Functional in water
- Maximum sample rate of 100 S/s
It supports experiments that last between one second and 31 days.
: The following items are needed but not included: a battery module, viewer module (not needed if connecting to a computer to display data), USB module (not needed if using as a stand-alone probe or you have the Wi-Fi module), and Wi-Fi module (not needed if connecting to a computer through the USB module). A battery module and a USB module are recommended.
: For educational use only.
- Resolution:0.1 °C
ADC: 12 bit - Description:NeuLog surface temperature logger sensor
- Range:−40...+140 °C
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



