Ubbelohde ASTM Series Capillary Viscometers, Fungilab
Ubbelohde viscometers are used to determine kinematic viscosities of transparent Newtonian liquids such as jet and hydraulic lubricants.
: UNE 400313, ASTM D445-446, ASTM D2515 and ISO 3104-3105).
: Includes constant value calibration certificate at 40ºC (104ºF) and 100ºC (212ºF).
Frequently Bought Together