Order Entry
Mexico
ContactUsLinkComponent
Aluminum ICP Standard, 1 ml = 1 mg Al (1000 ppm Al)Al in 3% HCl, Ricca Chemical Company
Aluminum ICP Standard, 1 ml = 1 mg Al (1000 ppm Al)Al in 3% HCl, Ricca Chemical Company
Catalog # RCPAL1KH-500
Supplier:  RICCA CHEMICAL CO
Small Business Enterprise
CAS Number:  
undefined
Aluminum ICP Standard, 1 ml = 1 mg Al (1000 ppm Al)Al in 3% HCl, Ricca Chemical Company
Catalog # RCPAL1KH-500
Supplier:  RICCA CHEMICAL CO
Supplier Number:  RPAL1KH0-500N
Small Business Enterprise
CAS Number:  

Specifications

  • Concentration:
    1000 µg/ml Al
  • Matrix:
    3% HCl
  • Size:
    500 ml
  • Cat. no.:
    RCPAL1KH-500
  • Element:
    Aluminum
  • Supplier No.:
    RPAL1KH0-500N

Specifications

About this item