Ammonium Standard, 1 ml = 1 mg NH₄+, 1000 ppm NH₄+ (776.5 ppm N), Ricca Chemical Company
Catalog # RC6934
Proveedor: RICCA CHEMICAL CO
Small Business Enterprise
Número CAS:
Especificaciones
- Concentration:1000 ppm
- Pack type:Plastic Bottle
- Description:Ammonium standard solution
- Size:120 ml
- Cat. no.:RC6934
- Contents:NH₄Cl in H₂O
- Supplier No.:R0693000-120A