Linear Alkylbenzene Sulfonate (LAS) Standard, 1 ml = 0.01 mg LAS, 10 ppm LAS, Ricca Chemical Company
Catalog # RC434816
Supplier: RICCA CHEMICAL CO
Small Business Enterprise
CAS Number:
Specifications
- Size:500 ml
- Cat. no.:RC434816
- Supplier No.:R4348000-500C