Order Entry
Mexico
ContactUsLinkComponent
analytical reagent
Health
Irritant
analytical reagent
Catalog # RC4124-32
Supplier:  RICCA CHEMICAL CO
Small Business Enterprise
CAS Number:  
undefined
Health
Irritant
analytical reagent
Catalog # RC4124-32
Supplier:  RICCA CHEMICAL CO
Supplier Number:  4124-32
Small Business Enterprise
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Size:
    1 L
  • Cat. no.:
    RC4124-32
  • Supplier No.:
    4124-32

Specifications

Specification Test Results

  • Appearance:
    Clear, dark brown, Iodine odor
  • Assay at 20 °C (traceable to NIST SRM 136):
    0.1576 ± 0.0002 N at 20.0 °C

Specification Test Results

About this item

 :
Boiling Pt:~ 100 °C
Melting Pt:~ 0 °C
Density:1.0400 g/cm³
 :GHS08
 :GHS07
Storage Temperature:Ambient