Order Entry
Mexico
Orders LinkContactUsLinkComponent
Visual Scientifics Base
Visual Scientifics Base
Visual scientifics base, 25×16×2.5"
Catalog # 470148-822
Supplier #:  PTBASE
Visual Scientifics Base
Catalog # 470148-822
Visual Scientifics Base
Visual Scientifics Base
Visual scientifics base, 25×16×2.5"
Clearance
$139.50
Sale
Valid until 31/12/2026
Log in for availability
Each (1UN) Case de 20 (1UN)
Product Details & Documents
This base platform supports the full range of Visual Scientifics activities and accessories.
  • High-contrast grid
  • Magnetic
  • Platform neutral - works with your preferred probeware/video analysis tools
Your students can set up their experiments quickly and easily with this large, sturdy, magnetic base that features threaded junction points for precise probeware mounting. Used in conjunction with Visual Scientifics accessory equipment, this laboratory investigation system offers students an engaging platform and accurate results. Its high-contrast color grid also allows for easy data collection with video analysis systems.
Specifications
  • Description:
    Visual scientifics base
  • Size:
    25×16×2.5"
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Visual scientifics wave demonstration
    Visual scientifics wave demonstration
    470212-494
    Log in for availability
    Each (1UN) - $1,498.82