Order Entry
Mexico
ContactUsLinkComponent
Calcium sulphate with indicator (Cobalt (II) chloride)
Health
Irritant
Environment
Calcium sulphate with indicator (Cobalt (II) chloride)
Catalog # JTL057-7
CAS Number:  
MOC:  Non-RegulatedCertificates
undefined
Health
Irritant
Environment
Calcium sulphate with indicator (Cobalt (II) chloride)
Catalog # JTL057-7
Supplier Number:  L057-07
CAS Number:  
MOC:  Non-RegulatedCertificates

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Poly bottle
  • Size:
    454 g
  • Cat. no.:
    JTL057-7
  • Supplier No.:
    L057-07

Specifications

About this item

 :
 :136.14 g/mol
Storage Temperature:Ambient
EINECS:231-900-3
UN:3077
ADR:9,III
 :GHS08
 :GHS07
 :GHS09
CAS Number:7778-18-9
Formula:CaO₄S
MDL Number:MFCD00010912