Order Entry
Mexico
ContactUsLinkComponent
Potassium hexacyanoferrate (III) ≥99%, crystals BAKER ANALYZED® ACS
Potassium hexacyanoferrate (III) ≥99%, crystals BAKER ANALYZED® ACS
Catalog # JT3104-1
CAS Number:  
MOC:  Non-RegulatedCertificates
undefined
Potassium hexacyanoferrate (III) ≥99%, crystals BAKER ANALYZED® ACS
Catalog # JT3104-1
Supplier Number:  3104-01
CAS Number:  
MOC:  Non-RegulatedCertificates

Specifications

  • Pack type:
    Glass bottle, amber, wide mouth
  • Size:
    500 g
  • Cat. no.:
    JT3104-1
  • Supplier No.:
    3104-01

Specifications

Specification Test Results

  • Appearance:
    Passes Test
  • Assay (K₃Fe(CN)₆) (by iodometry):
    ≥99.0 %
  • Insoluble Matter:
    ≤0.005 %
  • Chloride (Cl):
    ≤0.01 %
  • Sulfate (SO₄):
    ≤0.01 %
  • Ferro Compounds (as [Fe(CN)₆]⁴⁻):
    ≤0.05 %

Specification Test Results

About this item

CAS Number:13746-66-2
 :
Formula:K₃Fe(CN)₆
MDL Number:MFCD00011392
 :329.25 g/mol
Merck Index:13,07712
Density:1.85 g/cm³ (20 °C)
 :Management of Change (MOC) category = N
Storage Temperature:Ambient
EINECS:237-323-3