

Lead (II) acetate trihydrate ≥99%, granules BAKER ANALYZED® ACS
Catalog # JT2271-1
Supplier: Avantor
CAS Number:
MOC: Non-RegulatedCertificates
Specifications
- Pack type:Poly bottle
- Size:500 g
- Cat. no.:JT2271-1
- Supplier No.:2271-01
Specifications
Specification Test Results
- Assay ((CH₃COO)₂Pb·3H₂O) (by EDTA titrn):99.0 - 103.0 %
- Insoluble Matter:≤0.01 %
- pH of 5% Solution at 25°C:5.5 - 6.5
- Nitrate and Nitrite (as NO₃):≤0.005 %
- Calcium (Ca):≤0.005 %
- Copper (Cu):≤0.002 %
- Trace Impurities - Iron (Fe):≤0.001 %
- Potassium (K):≤0.005 %
- Sodium (Na):≤0.010 %
- Chloride (Cl):≤5 ppm
Specification Test Results
About this item
:
:379.34 g/mol
Merck Index:13,05415
Boiling Pt:280 °C (1013 hPa)
Boiling Pt:280 °C (1013 hPa)
Melting Pt:75 °C
Density:2.55 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:206-104-4
UN:1616
ADR:6.1,III
:Management of Change (MOC) category = N
:GHS08
:GHS09
CAS Number:6080-56-4
Formula:(H₃CCOO)₂Pb·3H₂O
MDL Number:MFCD00150023