Ammonium thiocyanate, crystals BAKER ANALYZED® ACS
JT0818-1
: Avantor
:
- Pack type:Poly bottle, white, blue T/E cap
- Size:500 g
- Cat. no.:JT0818-1
- Supplier No.:0818-01
- Appearance:Passes Test
- Assay (NH₄SCN) (by Ag titrn):≥97.5 %
- Insoluble Matter:≤0.005 %
- Residue after Ignition:≤0.010 %
- pH of 5% Solution at 25°C:4.5 - 6.0
- Chloride (Cl):≤0.005 %
- Sulfate (SO₄):≤0.005 %
- Iodine Consuming Substances (meq/g):≤0.004
- Trace Impurities - ACS - Heavy Metals (as Pb):≤5 ppm
- Trace Impurities - Iron (Fe):≤3.000 ppm
:
:76.12 g/mol
:13,00564
:150 °C
:1.305 g/cm³ (20 °C)
:Management of Change (MOC) category = NI
:217-175-6
:GHS07
:Ambient
:1762-95-4
:NH₄SCN
:MFCD00011428