
Citric acid monohydrate ≥99.5%, granules BP|Ph. Eur.|JP|USP
Catalog # JT0116-7
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
CAS Number:
Specifications
- Pack type:PE pail, screw top
- Size:12 kg
- Cat. no.:JT0116-7
- Supplier No.:0116-07
Specifications
Specification Test Results
- USP - Assay (C₆H₈O₇) (anhydrous basis):99.5 - 100.5 %
- USP - Clarity of Solution:Passes Test
- USP - Color of Solution:Passes Test
- USP - Identification:Passes Test
- USP - Oxalic Acid:≤0.036 %
- USP - Readily Carbonizable Substances:Passes Test
- USP - Residue on Ignition:≤0.1 %
- USP - Sulfate (SO₄):≤0.015 %
- USP - Water (H₂O):7.5 - 9.0 %
- EP/BP - Assay (C₆H₈O₇) (anhydrous basis):99.5 - 100.5 %
- EP/BP - Appearance of Solution:Passes Test
- EP/BP - Ash (sulfated):≤0.1 %
- EP/BP - Endotoxin Concentration, IU/mg:≤0.5
- EP/BP - Identification B:Passes Test
- EP/BP - Identification E:Passes Test
- EP/BP - Oxalic Acid:≤360 ppm
- EP/BP - Readily Carbonizable Substances:Passes Test
- EP/BP - Sulfate (SO₄):≤150 ppm
- EP/BP - Water (H₂O):7.5 - 9.0 %
- JP - Assay (HOC(COOH)(CH₂COOH)₂) (anhydrous basis):99.5 - 100.5 %
- JP - Clarity and Color of Solution:Passes Test
- JP - Heavy Metals (as Pb):≤10 ppm
- JP - Identification:Passes Test
- JP - Oxalic Acid:≤360 ppm
- JP - Water (H₂O):7.5 - 9.0 %
- JP - Readily Carbonizable Substances:Passes Test
- JP - Residue on Ignition:≤0.10 %
- JP - Sulfate (SO₄):≤150 ppm
- ACS - Iron (Fe):≤3 ppm
- Lead (Pb):≤2 ppm
Specification Test Results
About this item
:
:210.14 g/mol
Merck Index:13,02350
Melting Pt:135…152 °C
Density:1.552 g/cm³ (20 °C)
:Management of Change (MOC) category = R
Flash Pt:173.9 °C (closed cup)
EINECS:201-069-1
:GHS07
Storage Temperature:Ambient
CAS Number:5949-29-1
Formula:C₆H₈O₇.H₂O
MDL Number:MFCD00149972
Melting Pt:135 - 152 °C