ESK® Sampling Kit, with Tube, Swab and Letheen Broth, Puritan Medical Products
Catalog # 89221-708
Supplier: PURITAN MEDICAL PRODUCTS COMP
Product available on GSA Advantage®
Specifications
- Sterility:Sterile
- Environmentally Preferable:
- Cat. no.:89221-708
- Tip Size:17.4 mm (0.687")
- Handle Length:82.55 mm (3.25")
- Tip Diameter:5.15 mm (0.203")
- Overall Length:103.12 mm (4.06") (tube)
- Fill Solution:10 mL Letheen Broth
- Supplier No.:25-83010-PDBLB
Specifications
About this item
Easy-to-use, reliable test kits are available with Neutralizing Buffer, Buffered Peptone Water, Letheen Broth, or Butterfields Solution, in a 4mL or 10mL fill
Letheen Broth is recommended for the neutralization of quaternary ammonium compounds in disinfectant testing.
Pre-filled with 4mL or 10mL Letheen Broth. Shelf life is 18 months. Made in USA.
They come with a small polyester tip attached to a quick-turn, leak-resistant cap to ensure optimum control.
Ordering information: Kits are packaged 50 individually wrapped tubes per case. Bulk packaging includes 6 boxes of 50 individually wrapped tubes per box.