Order Entry
Mexico
ContactUsLinkComponent
Metal Stopcocks, Cadence Science®
  75997-206
undefined
Metal Stopcocks, Cadence Science®
  75997-206
 :  6019IND
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
  • Description:
    1-Way Stopcocks with 1 Female Luer Lock and 1 Male Luer Lock
  • Material:
    Plated brass
  • Cat. no.:
    75997-206
Made from either nickel plated brass or stainless steel, Cadence Science® metal stopcocks are precision-machined, individually lapped, and carefully mated to maintain pressure characteristics
All stopcocks are pressure resistant to 45 psi except the high pressure variants which are resistant up to 290 psi (20 bars). The spring clip stopcocks are designed to hold the key in position under proper sealing pressure and do not require a screwdriver in cleaning operations. Standard luer tapers include Female Luer Lock (FLL), Male Luer Lock (MLL), and Male Luer (ML).