Order Entry
Mexico
ContactUsLinkComponent
Corning® Modified Impeller Assembly for Magnetically-Driven Corning® Proculture® Spinner Flask, Corning
  89089-950
 :  Corning
Product available on GSA Advantage®
undefined
Corning® Modified Impeller Assembly for Magnetically-Driven Corning® Proculture® Spinner Flask, Corning
  89089-950
 :  Corning
 :  401901
Product available on GSA Advantage®
  • Description:
    Modified Impeller Assembly for Magnetically-Driven 36L Corning® Proculture® Spinner Flask, Reusable
  • Cat. no.:
    89089-950
  • Fits Flask:
    36L Corning® Proculture® spinner flask (22877-088, 22877-098, 22877-108, 22877-132, 22877-138, 22877-142)
  • Supplier No.:
    401901
Stainless steel impeller shaft with dual bearing, modified impeller assembly for use in magnetically driven, Proculture® Spinner Flasks
  • Narrower impeller paddle design allows use with probes to create a small bioreactor
  • Non sterile