Ethanol absolute ≥99.5% (v/v) USP for biochemistry
71006-012
:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Pack type:Glass bottle, clear
- Size:475 ml (16 oz.)
- Cat. no.:71006-012
- Supplier No.:3916EA
- RNA-ase Negative:Pass
- DNA-ase Negative:Pass
- Assay 200 proof:99.5 (% v/v) Ethanol
- Water:0.1
- Specific Gravity @15.56˚C (USP):0.7962
- Specific Gravity @15.0˚C (JP):0.797
- Relative Density @ 20˚C (BP/EP):0.790-0.793
- Color:Passes USP test for color
- Clarity:Passes USP Test for Clarity
- Acidity:30 ppm
- Alkalinity:Passes USP Test
- Non Volatile Residue:25 ppm
- Sum of Acetal &Acetaldehyde:10 ppm
- Benzene:2 ppm
- Methanol:100 ppm
- UV Absorbance @ 240 nm:0.40 A (USP/BP/EP/JP)
- UV Absorbance @ 250nm:0.30 A (USP/BP/EP/JP)
- UV Absorbance @ 260nm:0.30 A (USP/BP/EP/JP)
- UV Absorbance @ 270:0.10 A (USP/BP/EP/JP)
- UV Absorbance @ 235- 340 nm:Pass (USP/BP/EP/JP)
:
:46.07 g/mol
:12,03806
:78.3 °C (1013 hPa)
:78.3 °C (1013 hPa)
:–117 °C
:-117 °C
:0.7895 g/cm³ (20 °C)
:12 °C
:12 °C
:Ambient
:200-578-6
:1170
:3,II
:01-2119457610-43
:GHS02
:64-17-5
:C₂H₅OH
:MFCD00003568