Order Entry
Mexico
ContactUsLinkComponent
Accessories for Barnstead Pacific TII Water Purification Systems, Thermo Scientific
Accessories for Barnstead Pacific TII Water Purification Systems, Thermo Scientific
Catalog # 89230-312
Product available on GSA Advantage®
Accessories for Barnstead Pacific TII Water Purification Systems, Thermo Scientific
Catalog # 89230-312
Supplier Number:  09.4000
Product available on GSA Advantage®
Specifications
  • Description:
    Double Pretreatment System with 2 10" housings, combi cartridge (5 micron filter with activated carbon), and hardness stabilizer cartridge
  • Cat. no.:
    89230-312
  • For Use With:
    All Pacific TII Systems
  • Supplier No.:
    09.4000