Easy-Read® Thermometers feature Enviro-Safe liquid- the only EnviroKleen™ certified environmentally friendly thermometer liquid available on the market. Enviro-Safe liquid is dyed black and put against yellow back glass creating a high contrast and making this thermometer easy-to-read. Each Thermometer is individually serialized and Includes a multi-language (English, French, German, Italian, Portuguese and Spanish) Statement of Accuracy indicating accuracy traceable to NIST and DKD/PTB. Thermometers are accuracy to the requirements of ANSI/SAMA Z236.1-1983, ASTM, NIST, DKD and/or equivalent standards. Each item is tested and calibrated in H-B's exclusive triple accredited/registered ISO/IEC 17025:2005, A2LA accredited laboratory against equipment whose calibration is traceable to NIST and DKD/PTB. Individually packaged in Safety-Pak™ packaging to prevent breakage. Glass color: Yellow.
- Non-toxic and biodegradable liquid
- Lead-free
- Yellow back glass
- Easy to read
Ideal for use in chemical/petrochemical laboratories, universities, scientific institutions, food and beverage processing, pharmaceutical and medical industries, dairies, breweries and distilleries.
- Catalog No:
- 89217-924
- 89217-922
- Environmentally Preferable:
- Safer Human and Environmental Health
- Safer Human and Environmental Health