Wide Range Hydrion Double Roll Dispenser
Catalog # 470144-894
Supplier: MICRO ESSENTIAL LABORATORY I
Specifications
- Description:Wide Range Hydrion Double Roll Dispenser
- Cat. no.:470144-894
- pH Range:pH 1 - 12
- Supplier No.:140
Specifications
About this item
See Dramatic Color Changes
- Two 4.6 m rolls in one dispenser
- Tests from 1-12 on pH scale in 1.0 units
- Color chart included