Order Entry
Mexico
ContactUsLinkComponent
Vorticella Slide
Vorticella Slide
  470177-506
 :  Avantor
undefined
Vorticella Slide
  470177-506
 :  Avantor
  • Description:
    Vorticella Slide
  • Cat. no.:
    470177-506
  • Supplier No.:
    WARD470177-506
  • Orientation:
    Whole mount
Species of protozoa
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Peritrichous ciliate, contractile stalk and band-shaped nucleus.