Order Entry
Mexico
Orders LinkContactUsLinkComponent
Volta's Hail Storm
Volta's Hail Storm
Volta's Hail Storm
Catalog # 470200-214
Supplier #:  N-132
Volta's Hail Storm
Catalog # 470200-214
Volta's Hail Storm
Volta's Hail Storm
Volta's Hail Storm
Price
$2,867.05
Log in for availability
Each (1UN)
Product Details & Documents
A Perfect accessory for the Electrostatic Generator
  • Simple to use
  • Attaches to any Van De Graaff
Place pith balls or bits of aluminum foil in this cylinder to see how particles are attracted to the upper plate by induction, then repelled upon contact.
Specifications
  • Description:
    Volta's Hail Storm