Variable Gap Magnet
Catalog # 470121-488
Supplier: ELECTRONIC INSTRUMENTATION
Specifications
- Description:Variable Gap Magnet
- Cat. no.:470121-488
- Supplier No.:PH0801VGM
Specifications
About this item
Adjust the Magnets According To Your Needs.
- This apparatus consists of two neodymium magnets set on screws on a metal base
- Versatile and customizable, the magnet positions can be altered between 3 mm and 83 mm or removed to implement other magnets
- A pair of metal plates are included to provide a uniform magnetic field when performing experiments