Vacuum Chamber
Vacuum Chamber, H x OD, in.: 6-5/8 x 9-3/8
Catalog # 470211-850
Supplier: Thermo Fisher Scientific
Supplier #: 5305-0609
Jump to:
Product Details & Documents
Nalgene Complete polyetherimide jar and vacuum plate chamber provides strength and good chemical resistance.
Neoprene gasket provides secure seal. Slight amber cast of jar allows complete visibility of interior.
Plate has a center hole for exhaust with tube leading to an external hose connector which accepts 1/4" ID tubing.
For use at room temperature and at full vacuum.
Specifications
- Description:Vacuum Chamber, H x OD, in.: 6-5/8 x 9-3/8