Order Entry
Mexico
Orders LinkContactUsLinkComponent
 

USDA Soil Texturing Field Flow Chart

USDA Soil Texturing Field Flow Chart, 8.5", 11"
  470018-592
 :  USDA FC
USDA Soil Texturing Field Flow Chart
  470018-592
USDA Soil Texturing Field Flow Chart
USDA Soil Texturing Field Flow Chart

USDA Soil Texturing Field Flow Chart, 8.5", 11"

 
$532.49
   
Each (1UN)
 

Preparation for Work in the Field

For assisting with soil texturing procedures and assigning a texture class using the USDA texture triangle, this chart uses step-wise procedures for estimating sand, silt, and clay content. The ribbon method is implemented to estimate the fine-grained fraction followed by placing the fraction within the texture triangle to determine Texture Class (ex: sandy loam). The accompanying guide provides instruction for determining subclass (ex: fine sandy loam) and assigning textural class modifiers for rock fragments greater than 2mm (ex: gravelly silt loam) and organic compositional textural modifiers (ex: peaty sandy clay loam). Guide is printed on durable, double-sided, plastic-coated paper.

Size: 8 1/2" x 11".

 
  • Width:
    11"
  • Description:
    USDA Soil Texturing Field Flow Chart
  • Length:
    8.5"