Order Entry
Mexico
Orders LinkContactUsLinkComponent
Kemtec® Urinalysis
Kemtec® Urinalysis
Urinalysis Kit
Catalog # 470111-836
Supplier #:  8-306
Kemtec® Urinalysis
Catalog # 470111-836
Kemtec® Urinalysis
Kemtec® Urinalysis
Urinalysis Kit
Price
$7,648.66
Log in for availability
Each (1UN)
Product Details & Documents
Simulated urine testing.
  • No real or human products
  • Learn basic testing procedure
Urinalysis introduces students to to the world of biomedical technology. Using simulated samples, students evaluate the general characteristics of urine, observe for sediments, and test for glucose, albumin, pH, specific gravity, and lead.
Delivery information: See datasheet for contents information.
Documents
Specifications
  • Description:
    Urinalysis Kit