Urethra, Male Slide
470182-946
: Avantor
- Description:Urethra, Male
- Cat. no.:470182-946
- Stain:Hematoxylin and Eosin
- Supplier No.:WARD470182-946
- Orientation:Section
Conducts Fluids from the Body to the Outside Environment
- Identifying features clearly distinguishable
- Rigorous quality control standards
Mammal. Sectioned through prostate and portion of urethra.