Umbilical Cord Slide
Catalog # 470182-974
Supplier: Avantor
Specifications
- Description:Umbilical Cord
- Cat. no.:470182-974
- Stain:Hematoxylin and Eosin
- Supplier No.:WARD470182-974
- Orientation:Section
Specifications
About this item
Link Between Fetus and Placenta in Mammals During Development
- Identifying features clearly distinguishable
- Rigorous quality control standards
Human. Mucous tissue shows large intercellular spaces filled with Wharton’s jelly.