Tuning Fork and Resonator Box Set
Catalog # 470111-232
Supplier: UNITED SCIENTIFIC SUPPLIES
Specifications
- Description:Tuning Fork and Resonator Box Set
- Cat. no.:470111-232
- Supplier No.:TFRB256
Specifications
About this item
Produces a Louder Tone With a Greater Range of Audible Overtones
- Tuning fork is securely mounted on top
- Open end box increase the sound of the resonance
A physical pitch tuning fork has been specifically modified to screw into a resonator case. The unit consists of a tuning fork, wooden resonator box and a wooden wedge snapper for exciting the fork.