Order Entry
Mexico
ContactUsLinkComponent
Tree Finder
Tree Finder
Catalog # 470100-820
Supplier:  ADVENTUREKEEN
undefined
Tree Finder
Catalog # 470100-820
Supplier:  ADVENTUREKEEN
Supplier Number:  0912550015

Specifications

  • Description:
    Tree Finder
  • Length:
    14 cm
  • Cat. no.:
    470100-820
  • No. of Pages:
    58
  • Width:
    10 cm
  • Supplier No.:
    0912550015

Specifications

About this item

The classic key to identifying trees by their leaves and needles.

Covers native and commonly introduced trees of the U.S. and Canada east of the Rockies and north of Florida. Line drawings point out important features of plant structure.