Thymus, Mammal Slide
470182-782
: Avantor
- Description:Thymus, Mammal
- Cat. no.:470182-782
- Stain:Hematoxylin and Eosin
- Supplier No.:WARD470182-782
- Orientation:Section
Immune System Organ
- Identifying features clearly distinguishable
- Rigorous quality control standards
Mammal. Gland in stage of involution. Important in the development of T cells.