Order Entry
Mexico
Orders LinkContactUsLinkComponent
Ward's® This Old Bone Lab Activity
Ward's® This Old Bone Lab Activity
WARD’S This Old Bone Lab Activity, 6
Catalog # 470024-250
Supplier:   Avantor
Ward's® This Old Bone Lab Activity
Catalog # 470024-250
Supplier:  Avantor
Ward's® This Old Bone Lab Activity
Ward's® This Old Bone Lab Activity
WARD’S This Old Bone Lab Activity, 6
Price
$2,211.00
Log in for availability
Each (1UN)
Product Details & Documents
With This Old Bone Lab Activity, students perform an actual technique used by forensic anthropologists to determine the age of individuals at their time of death.
  • For Grades 10-12
  • Students Determine Age from Osteon Counts
  • Estimated Class Time Required: 45 Minutes
  • Teacher's Guide Included
They gain valuable microscopy and anatomy skills in this activity as they learn to count, measure, and analyze the Haversian systems in bone using a compound microscope with an ocular micrometer (both sold separately).
The lab requires 45 minutes.
 : This kit includes a stage micrometer, six prepared microscope slides of ground human bone, a teacher’s guide, and student copy masters. An ocular micrometer (reticle), a compound microscope, and adhesive tape are required but not included.
Specifications
  • Description:
    WARD’S This Old Bone Lab Activity
  • No. of Lab Groups:
    6
Accessories
Product Information
  • Catalog #
    Illumination
    Availability
    Price
  • Monocular
    Monocular
    470016-038
    LED corded
    Log in for availability
    Each (1UN) - $6,929.11