The World’s Simplest Motor™ Kit
470020-630
: Avantor
- Description:The World’s Simplest Motor™
- Environmentally Preferable:
- Cat. no.:470020-630
- Supplier No.:YEANY EDUCATIONAL PRODUCTS SE
Demonstrate the Fundamentals of Motor Operation
Electricity runs through the coil, forming a magnetic field; the coil is then “pushed” away from the magnet, causing the coil to spin and become charged. It includes assembly instructions and suggested activities. Size: 2 5/8"L x 1 3/8"W x 1 1/2"H.