Order Entry
Mexico
ContactUsLinkComponent
Testis, Spermatogenesis, Mammal Slide
Testis, Spermatogenesis, Mammal Slide
Catalog # 470177-626
Supplier:  Avantor
undefined
Testis, Spermatogenesis, Mammal Slide
Catalog # 470177-626
Supplier:  Avantor
Specifications
  • Description:
    Testis, Spermatogenesis, Mammal Slide
  • Environmentally Preferable:
  • Cat. no.:
    470177-626
  • Stain:
    Iron Hematoxylin
  • Supplier No.:
    WARD470177-626
  • Orientation:
    Section
Specifications
About this item
Male reproductive components
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Rodent. Shows capsule, seminiferous tubules, spermatogenic cells, sertoli, and Leydig’s cells.