Ten Specimen Density Set
Ten Specimen Density Set
Catalog # 470149-490
Supplier: VWR International
Jump to:
Product Details & Documents
This set features a handy, economical selection of density cubes.
- Economical
- Handy
- Cubes Measure 1" Each
- Materials Include Pine, Poplar, Plastic, and More
- Ten Density Cubes per Set
The cubes measure 1" each.
They're made of acrylic plastic, aluminum, brass, copper, nylon, oak, pine, poplar, PVC, and steel.
Specifications
- Description:Ten Specimen Density Set