Order Entry
Mexico
ContactUsLinkComponent
 
Systems Of The Human Body Chart Set
  470023-464
undefined
Systems Of The Human Body Chart Set
  470023-464
 :  03PS121L

 

  • Width:
    46 cm
  • Description:
    Human Body Set
  • Length:
    61 cm
  • Cat. no.:
    470023-464
  • Finish:
    Laminated
  • Supplier No.:
    03PS121L

 

 

Economical Set For An Overview Of The Most Studied Systems

  • Digestive, circulatory, nervous, and respiratory systems are detailed in this series

Each of the four charts includes labeled illustrations of that particular system and its components, detailed close-ups of key organs and processes, and text describing the main functions of the system and its parts.