Order Entry
Mexico
Orders LinkContactUsLinkComponent
Syringa, Leaf Slide
Syringa, Leaf Slide
Syringa, Leaf Slide, Cross section, Quadruple
Catalog # 470176-892
Supplier:   Avantor
Syringa, Leaf Slide
Catalog # 470176-892
Supplier:  Avantor
Syringa, Leaf Slide
Syringa, Leaf Slide
Syringa, Leaf Slide, Cross section, Quadruple
Price
$197.00
Log in for availability
Each (1UN)
Product Details & Documents
Illustrates characteristics of a temperate, dicot leaf
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Lilac. A typical mesophytic dicot leaf.
Specifications
  • Description:
    Syringa, Leaf Slide
  • Stain:
    Quadruple
  • Orientation:
    Cross section