Super Sunprint™ Kit
Refill, Super Sunprint Kit, 15 Sheets, 20 cm x 25 cm each
Catalog # 470106-172
Supplier: UNIVERSITY OF CALIFORNIA
: W440B
Jump to:
Product Details & Documents
Make photographic prints using the sun.
To make images, place an array of interesting objects on a sheet of large Sunprint™ paper, expose it to the sun for a few minutes, immerse in water, and watch as a permanent image appears.
: Includes 15 sheets of sun-sensitive paper (20×30cm), an acrylic plate, and complete instructions.
: Use only as directed. Adult supervision recommended.
Specifications
- Description:Refill, Super Sunprint Kit, 15 Sheets, 20 cm x 25 cm each