Sublingual Gland, Mammal Slide
470182-834
: Avantor
- Description:Sublingual Gland, Mammal
- Cat. no.:470182-834
- Stain:Hematoxylin and Eosin
- Supplier No.:WARD470182-834
- Orientation:Section
Major Salivary Gland
- Identifying features clearly distinguishable
- Rigorous quality control standards
Mammal. A mixed gland showing mucous alveoli and ducts with scattered serous crescents, or demilunes.