Student Magnetic Field Demonstrator
470110-162
- Description:Student Magnetic Field Demonstrator
- Cat. no.:470110-162
- Supplier No.:MGFD01
Economical Illustration
- Size: 90 x 65 x 6mm
Iron filings sealed in a flat, transparent plastic case allow students to visualize the field of a small magnet without the usual mess.