Order Entry
Mexico
ContactUsLinkComponent
Stomach, Fundic Region, Mammal Slide
Catalog # 470012-776
Supplier:  Avantor
undefined
Stomach, Fundic Region, Mammal Slide
Catalog # 470012-776
Supplier:  Avantor
Specifications
  • Description:
    Stomach, Fundic Region, Mammal Slide
  • Cat. no.:
    470012-776
  • Stain:
    Hematoxylin and Eosin
  • Supplier No.:
    WARD470012-776
  • Orientation:
    Cross section
Specifications
About this item
Digestive organ
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Mammal. Simple tubular gastric glands contain a mixed population of cells: mucus-secreting; parietal, or hydrochloric acid; and zymogenic, or pepsin-secreting.