Stomach and Duodenum Slide
Stomach, & Duodenum, Longitudinal section, Hematoxylin and Eosin
Catalog # 470182-862
Supplier: Avantor
Jump to:
Product Details & Documents
Digestive System
- Identifying features clearly distinguishable
- Rigorous quality control standards
Mammal. Typical sections through the zone of plica circulares and pyloric sphincter.
Specifications
- Description:Stomach, & Duodenum
- Stain:Hematoxylin and Eosin
- Orientation:Longitudinal section